Merck
  • Preparation, characterization, and performance evaluation of UiO-66 analogues as stationary phase in HPLC for the separation of substituted benzenes and polycyclic aromatic hydrocarbons.
Preparation, characterization, and performance evaluation of UiO-66 analogues as stationary phase in HPLC for the separation of substituted benzenes and polycyclic aromatic hydrocarbons.
PloS one (2017-06-06)
Weiwei Zhao, Chaoyan Zhang, Zengguang Yan, Youya Zhou, Jianrong Li, Yabo Xie, Liping Bai, Lin Jiang, Fasheng Li
ABSTRACT
UiO-66 analogues are good candidates as stationary phase in HPLC because of their chemical/thermal stability, large surface area, and two cage structures. Here, two UiO-66 analogues, UiO-66-NH2 and UiO-67, were synthesized and used as stationary phase in HPLC to evaluate their performance in the separation of substituted benzenes (SBs) and polycyclic aromatic hydrocarbons (PAHs). The results showed that SBs could be well separated on UiO-66-NH2 column but not on UiO-67 column. Nonetheless, PAHs could be well separated on UiO-67 column. The separation mechanisms of SBs and PAHs on UiO-66 analogues may be involved in the pore size and functional group in the frameworks of UiO-66 analogues. Introduction of the-NH2 into UiO-66 significantly reduced its adsorption capacity for SB congeners, which resulted in less separation of SBs on UiO-66-NH2. As for the separation of PAHs on UiO-67 column, the π-π stacking effect was supposed to play a vital role.
Sigma-Aldrich
2-Aminoterephthalic acid, 99%