Merck
  • Cytotoxic effect of a mannogalactoglucan extracted from Agaricus bisporus on HepG2 cells.
Cytotoxic effect of a mannogalactoglucan extracted from Agaricus bisporus on HepG2 cells.
Carbohydrate polymers (2017-05-20)
Amanda do Rocio Andrade Pires, Andrea Caroline Ruthes, Silvia Maria Suter Correia Cadena, Marcello Iacomini
ABSTRACT
A mannogalactoglucan (RK2-Ab; M
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) bioluminescent somatic cell assay kit, for cellular ATP determination