Merck
  • Identification of a novel chalcone derivative that inhibits Notch signaling in T-cell acute lymphoblastic leukemia.
Identification of a novel chalcone derivative that inhibits Notch signaling in T-cell acute lymphoblastic leukemia.
Scientific reports (2017-05-21)
Mattia Mori, Luca Tottone, Deborah Quaglio, Nadezda Zhdanovskaya, Cinzia Ingallina, Marisa Fusto, Francesca Ghirga, Giovanna Peruzzi, Maria Elisa Crestoni, Fabrizio Simeoni, Francesca Giulimondi, Claudio Talora, Bruno Botta, Isabella Screpanti, Rocco Palermo
ABSTRACT
Notch signaling is considered a rational target in the therapy of several cancers, particularly those harbouring Notch gain of function mutations, including T-cell acute lymphoblastic leukemia (T-ALL). Although currently available Notch-blocking agents are showing anti-tumor activity in preclinical studies, they are not effective in all the patients and often cause severe side-effects, limiting their widespread therapeutic use. Here, by functional and biological analysis of the most representative molecules of an in house library of natural products, we have designed and synthetized the chalcone-derivative 8 possessing Notch inhibitory activity at low micro molar concentration in T-ALL cell lines. Structure-activity relationships were afforded for the chalcone scaffold. Short term treatments with compound 8 resulted in a dose-dependent decrease of Notch signaling activity, halted cell cycle progression and induced apoptosis, thus affecting leukemia cell growth. Taken together, our data indicate that 8 is a novel Notch inhibitor, candidate for further investigation and development as an additional therapeutic option against Notch-dependent cancers.
Sigma-Aldrich
GI254023X, ≥98% (HPLC)
Sigma-Aldrich
Galangin, autophagy inducing flavonoid
Sigma-Aldrich
γ-Secretase Inhibitor IX, Gamma-Secretase Inhibitor IX - CAS 208255-80-5, is a cell-permeable inhibitor of γ-secretase (Aβtotal IC₅₀ = 115 nM, Aβ42 IC₅₀ = 200 nM).