Merck
  • In vivo diagnostics of early abiotic plant stress response via Raman spectroscopy.
In vivo diagnostics of early abiotic plant stress response via Raman spectroscopy.
Proceedings of the National Academy of Sciences of the United States of America (2017-03-16)
Narangerel Altangerel, Gombojav O Ariunbold, Connor Gorman, Masfer H Alkahtani, Eli J Borrego, Dwight Bohlmeyer, Philip Hemmer, Michael V Kolomiets, Joshua S Yuan, Marlan O Scully
ABSTRACT
Development of a phenotyping platform capable of noninvasive biochemical sensing could offer researchers, breeders, and producers a tool for precise response detection. In particular, the ability to measure plant stress in vivo responses is becoming increasingly important. In this work, a Raman spectroscopic technique is developed for high-throughput stress phenotyping of plants. We show the early (within 48 h) in vivo detection of plant stress responses. Coleus (
Product Description
Supelco
Keracyanin chloride, analytical standard
Pricing and availability is not currently available.