Merck
  • X-34, a fluorescent derivative of Congo red: a novel histochemical stain for Alzheimer's disease pathology.
X-34, a fluorescent derivative of Congo red: a novel histochemical stain for Alzheimer's disease pathology.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2000-08-19)
S D Styren, R L Hamilton, G C Styren, W E Klunk
ABSTRACT
X-34, a lipophilic, highly fluorescent derivative of Congo red, was examined as a histochemical stain for pathological changes in Alzheimer's disease (AD). X-34 intensely stained neuritic and diffuse plaques, neurofibrillary tangles (NFTs), neuropil threads, and cerebrovascular amyloid. Comparison to standard methods of demonstrating AD pathology showed that X-34 correlated well with Bielschowsky and thioflavin-S staining. X-34 staining of NFTs correlated closely with anti-TAU antibody staining. A 1:1 correspondence of X-34 and anti-A beta antibody staining of plaques and cerebrovascular amyloid was observed. Both X-34 and thioflavin-S staining were eliminated by formic acid pretreatment, suggesting that beta-sheet secondary protein structure is a necessary determinant of staining. X-34 may be a general amyloid stain, like Congo red, because it also stains systemic amyloid deposits due to lambda-light chain monoclonal gammopathy. In conclusion, X-34 is a highly fluorescent marker for beta-sheet structures and intensely labels amyloid plaques, NFTs, neuropil threads, and vascular amyloid in AD brains. It can be used with both paraffin-embedded and frozen tissues as well as in combination with immunohistochemistry for double labeling. The intensity of staining and the simplicity and reproducibility of the technique suggest that it may be a useful addition to the standard techniques for evaluation of AD neuropathology. (J Histochem Cytochem 48:1223-1232, 2000)
Sigma-Aldrich
X-34, ≥90% (HPLC)
Pricing and availability is not currently available.