Merck
Foxa2 is required for the differentiation of pancreatic alpha-cells.
Developmental biology (2005-02-01)
Catherine S Lee, Newman J Sund, Rüdiger Behr, Pedro L Herrera, Klaus H Kaestner
ABSTRACT
The differentiation of insulin-producing beta-cells has been investigated in great detail; however, little is known about the factors that delineate the second-most abundant endocrine lineage, the glucagon-producing alpha-cell. Here we utilize a novel YAC-based Foxa3Cre transgene to delete the winged helix transcription factor Foxa2 (formerly HNF-3beta) in the pancreatic primordium during midgestation. The resulting Foxa2(loxP/loxP); Foxa3Cre mice are severely hypoglycemic and die within the first week of life. Mutant mice are hypoglucagonemic secondary to a 90% reduction of glucagon expression. While the number of mature glucagon-positive alpha-cells is dramatically reduced, specification of alpha-cell progenitors is not affected by Foxa2 deficiency. By marker gene analysis, we show that the expression of the alpha-cell transcription factors Arx, Pax6, and Brn4 does not require Foxa2 in the transcriptional hierarchy governing alpha-cell differentiation.