Merck
  • Strategies to Inhibit ABCB1- and ABCG2-Mediated Efflux Transport of Erlotinib at the Blood-Brain Barrier: A PET Study on Nonhuman Primates.
Strategies to Inhibit ABCB1- and ABCG2-Mediated Efflux Transport of Erlotinib at the Blood-Brain Barrier: A PET Study on Nonhuman Primates.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2016-08-06)
Nicolas Tournier, Sebastien Goutal, Sylvain Auvity, Alexander Traxl, Severin Mairinger, Thomas Wanek, Ourkia-Badia Helal, Irène Buvat, Michael Soussan, Fabien Caillé, Oliver Langer
ABSTRACT
The tyrosine kinase inhibitor erlotinib poorly penetrates the blood-brain barrier (BBB) because of efflux transport by P-glycoprotein (ABCB1) and breast cancer resistance protein (ABCG2), thereby limiting its utility in the treatment of non-small cell lung cancer metastases in the brain. Pharmacologic strategies to inhibit ABCB1/ABCG2-mediated efflux transport at the BBB have been successfully developed in rodents, but it remains unclear whether these can be translated to humans given the pronounced species differences in ABCG2/ABCB1 expression ratios at the BBB. We assessed the efficacy of two different ABCB1/ABCG2 inhibitors to enhance brain distribution of Papio anubis baboons underwent PET scans of the brain after intravenous injection of Under baseline conditions, We successfully implemented ABCB1/ABCG2 inhibition protocols in nonhuman primates resulting in pronounced increases in brain distribution of
Sigma-Aldrich
Erlotinib hydrochloride