Merck
  • TEMPO supported on magnetic C/Co-nanoparticles: a highly active and recyclable organocatalyst.
TEMPO supported on magnetic C/Co-nanoparticles: a highly active and recyclable organocatalyst.
Chemistry (Weinheim an der Bergstrasse, Germany) (2008-07-31)
Alexander Schätz, Robert N Grass, Wendelin J Stark, Oliver Reiser
ABSTRACT
TEMPO was grafted on graphene-coated nanobeads with a magnetic cobalt core by using a general applicable "click"-chemistry protocol. The new heterogeneous CoNP-TEMPO emerged as a highly active catalyst for the chemoselective oxidation of primary and secondary alcohols using bleach as terminal oxidant. The outstanding stability of the C/Co nanoparticles enables the nanopowder to tolerate several TEMPO-mediated iterative oxidation reactions without any significant loss in catalyst activity. Furthermore, the excellent magnetic properties enable the rapid separation and quantitative recycling of CoNP-TEMPO out of the reaction mixture by simple magnetic decantation. The recovered nanoparticles can be subsequently reused without any further purification.
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%