Merck
Substrate-targeting gamma-secretase modulators.
Nature (2008-06-13)
Thomas L Kukar, Thomas B Ladd, Maralyssa A Bann, Patrick C Fraering, Rajeshwar Narlawar, Ghulam M Maharvi, Brent Healy, Robert Chapman, Alfred T Welzel, Robert W Price, Brenda Moore, Vijayaraghavan Rangachari, Bernadette Cusack, Jason Eriksen, Karen Jansen-West, Christophe Verbeeck, Debra Yager, Christopher Eckman, Wenjuan Ye, Sarah Sagi, Barbara A Cottrell, Justin Torpey, Terrone L Rosenberry, Abdul Fauq, Michael S Wolfe, Boris Schmidt, Dominic M Walsh, Edward H Koo, Todd E Golde
ABSTRACT
Selective lowering of Abeta42 levels (the 42-residue isoform of the amyloid-beta peptide) with small-molecule gamma-secretase modulators (GSMs), such as some non-steroidal anti-inflammatory drugs, is a promising therapeutic approach for Alzheimer's disease. To identify the target of these agents we developed biotinylated photoactivatable GSMs. GSM photoprobes did not label the core proteins of the gamma-secretase complex, but instead labelled the beta-amyloid precursor protein (APP), APP carboxy-terminal fragments and amyloid-beta peptide in human neuroglioma H4 cells. Substrate labelling was competed by other GSMs, and labelling of an APP gamma-secretase substrate was more efficient than a Notch substrate. GSM interaction was localized to residues 28-36 of amyloid-beta, a region critical for aggregation. We also demonstrate that compounds known to interact with this region of amyloid-beta act as GSMs, and some GSMs alter the production of cell-derived amyloid-beta oligomers. Furthermore, mutation of the GSM binding site in the APP alters the sensitivity of the substrate to GSMs. These findings indicate that substrate targeting by GSMs mechanistically links two therapeutic actions: alteration in Abeta42 production and inhibition of amyloid-beta aggregation, which may synergistically reduce amyloid-beta deposition in Alzheimer's disease. These data also demonstrate the existence and feasibility of 'substrate targeting' by small-molecule effectors of proteolytic enzymes, which if generally applicable may significantly broaden the current notion of 'druggable' targets.
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Naproxen sodium, 98.0-102.0%