Merck
  • Scope and mechanism of enantioselective Michael additions of 1,3-dicarbonyl compounds to nitroalkenes catalyzed by nickel(II)-diamine complexes.
Scope and mechanism of enantioselective Michael additions of 1,3-dicarbonyl compounds to nitroalkenes catalyzed by nickel(II)-diamine complexes.
Journal of the American Chemical Society (2007-08-28)
David A Evans, Shizue Mito, Daniel Seidel
ABSTRACT
Readily prepared Ni(II)-bis[(R,R)-N,N'-dibenzylcyclohexane-1,2-diamine]Br(2) was shown to catalyze the Michael addition of 1,3-dicarbonyl compounds to nitroalkenes at room temperature in good yields with high enantioselectivities. The two diamine ligands in this system each play a distinct role: one serves as a chiral ligand to provide stereoinduction in the addition step while the other functions as a base for substrate enolization. Ligand modification within the catalyst was also investigated to facilitate the reaction of aliphatic nitroalkenes, 1,3-diketones, and beta-ketoacids. Ni(II)-bis[(R,R)-N,N'-di-p-bromo-benzylcyclohexane-1,2-diamine]Br(2) was found to be an effective catalyst in these instances. Furthermore, monodiamine complex, Ni(II)-[(R,R)-N,N'-dibenzylcyclohexane-1,2-diamine]Br(2), catalyzed the addition reaction in the presence of water. The proposed model for stereochemical induction is shown to be consistent with X-ray structure analysis.
Sigma-Aldrich
3,5-Heptanedione, 97%