Merck
  • Synthesis of indenes by the transition metal-mediated carboannulation of alkynes.
Synthesis of indenes by the transition metal-mediated carboannulation of alkynes.
The Journal of organic chemistry (2006-12-30)
Daohua Zhang, Zhijian Liu, Eul K Yum, Richard C Larock
ABSTRACT
The synthesis of highly substituted indenes has been achieved by three different transition metal-mediated methods. The first method involves the palladium-catalyzed carboannulation of internal alkynes. The second method utilizes a two-step approach, which involves first the palladium/copper-catalyzed cross-coupling of terminal alkynes with appropriately functionalized aryl halides, followed by copper-catalyzed intramolecular cyclization. The third method involves intermolecular palladium-catalyzed arylation of the arylalkynes formed in the first step of the second method.
Sigma-Aldrich
1-(Trimethylsilyl)propyne, 99%