Merck
A modular cross-linking approach for exploring protein interactions.
Journal of the American Chemical Society (2003-02-27)
Michelle Trester-Zedlitz, Katsuhiko Kamada, Stephen K Burley, David Fenyö, Brian T Chait, Tom W Muir
ABSTRACT
A method is described for the elucidation of protein-protein interactions using novel cross-linking reagents and mass spectrometry. The method incorporates (1) a modular solid-phase synthetic strategy for generating the cross-linking reagents, (2) enrichment and digestion of cross-linked proteins using microconcentrators, (3) mass spectrometric analysis of cross-linked peptides, and (4) comprehensive computational analysis of the cross-linking data. This integrated approach has been applied to the study of cross-linking between the components of the heterodimeric protein complex negative cofactor 2.
Sigma-Aldrich
N-Boc- 2,2′-(ethylenedioxy)diethylamine, ≥95.0% (NT)