Merck
  • N-isopropylidene-N'-2-nitrobenzenesulfonyl hydrazine, a reagent for reduction of alcohols via the corresponding monoalkyl diazenes.
N-isopropylidene-N'-2-nitrobenzenesulfonyl hydrazine, a reagent for reduction of alcohols via the corresponding monoalkyl diazenes.
The Journal of organic chemistry (2007-02-06)
Mohammad Movassaghi, Omar K Ahmad
ABSTRACT
The reagent N-isopropylidene-N'-2-nitrobenzenesulfonyl hydrazine (IPNBSH) is used in the reduction of alcohols via the loss of dinitrogen from transiently formed monoalkyl diazene intermediates accessed by sequential Mitsunobu displacement, hydrolysis, and fragmentation under mild reaction conditions.