Merck
Synthesis of ovalicin starting from D-mannose.
The Journal of organic chemistry (2005-11-19)
Shunya Takahashi, Nobuyuki Hishinuma, Hiroyuki Koshino, Tadashi Nakata
ABSTRACT
[reaction: see text] A new synthesis of epoxyketone 22 is described that is a key intermediate in Barton's synthesis of ovalicin (2), a powerful anti-angiogenetic inhibitor. The key process for the construction of 22 was ring-closing metathesis of olefins 11 and 12 obtained from 2,3:5,6-di-O-isopropylidene-alpha-D-mannofuranose (4) and regioselective desilylation of tri-TES ether 19. Furthermore, an alternative stereoselective route from 22 into 2 has also been developed, and the overall yield of 2 from 4 was 10.0%.
Sigma-Aldrich
2,3:5,6-Di-O-isopropylidene-α-D-mannofuranose, 97%
Pricing and availability is not currently available.