Merck
  • Epigenetic downregulation of the ISG15-conjugating enzyme UbcH8 impairs lipolysis and correlates with poor prognosis in nasopharyngeal carcinoma.
Epigenetic downregulation of the ISG15-conjugating enzyme UbcH8 impairs lipolysis and correlates with poor prognosis in nasopharyngeal carcinoma.
Oncotarget (2015-10-28)
Xiaoying Zhou, Jiazhang Wei, Fu Chen, Xue Xiao, Tingting Huang, Qian He, Shumin Wang, Chunping Du, Yingxi Mo, Longde Lin, Ying Xie, Lili Wei, Ying Lan, Mairiko Murata, Guangwu Huang, Ingemar Ernberg, Liudmila Matskova, Zhe Zhang
ABSTRACT
We identified the UBE2L6 gene, encoding the ISG15-conjugating enzyme UbcH8, as one gene significantly downregulated by promoter hypermethylation in nasopharyngeal carcinoma (NPC). Reduced expression of the UbcH8 protein correlated with poor outcome in NPC patients. Restored expression of UBE2L6 suppressed proliferation and colony formation in NPC cells, while inducing apoptosis. Of particular interest, we found that aberrant lipid turnover was controlled by UbcH8 in NPC through ISG15-conjugation of valosin-containing protein (VCP). Tumor tissue and NPC cell lines showed conspicuously strong accumulation of lipid droplets (LDs) compared to control nasopharyngeal epithelium and non-cancerous cell lines. We demonstrated that UbcH8 counteracts degradation of adipocyte triglyceride lipase (ATGL), a key enzyme in lipid catabolism.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone AA13, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human UBE2L6
Pricing and availability is not currently available.