Merck
  • VCIP135 acts as a deubiquitinating enzyme during p97-p47-mediated reassembly of mitotic Golgi fragments.
VCIP135 acts as a deubiquitinating enzyme during p97-p47-mediated reassembly of mitotic Golgi fragments.
The Journal of cell biology (2004-03-24)
Yanzhuang Wang, Ayano Satoh, Graham Warren, Hemmo H Meyer, Yangzhuang Wang
ABSTRACT
The AAA-ATPase p97/Cdc48 functions in different cellular pathways using distinct sets of adapters and other cofactors. Together with its adaptor Ufd1-Npl4, it extracts ubiquitylated substrates from the membrane for subsequent delivery to the proteasome during ER-associated degradation. Together with its adaptor p47, on the other hand, it regulates several membrane fusion events, including reassembly of Golgi cisternae after mitosis. The finding of a ubiquitin-binding domain in p47 raises the question as to whether the ubiquitin-proteasome system is also involved in membrane fusion events. Here, we show that p97-p47-mediated reassembly of Golgi cisternae requires ubiquitin, but is not dependent on proteasome-mediated proteolysis. Instead, it requires the deubiquitinating activity of one of its cofactors, VCIP135, which reverses a ubiquitylation event that occurs during mitotic disassembly. Together, these data reveal a cycle of ubiquitylation and deubiquitination that regulates Golgi membrane dynamics during mitosis. Furthermore, they represent the first evidence for a proteasome-independent function of p97/Cdc48.