Merck
  • Different apoptotic pathways activated by oxaliplatin in primary astrocytes vs. colo-rectal cancer cells.
Different apoptotic pathways activated by oxaliplatin in primary astrocytes vs. colo-rectal cancer cells.
International journal of molecular sciences (2015-03-12)
Matteo Zanardelli, Laura Micheli, Raffaella Nicolai, Paola Failli, Carla Ghelardini, Lorenzo Di Cesare Mannelli
ABSTRACT
Oxaliplatin-based chemotherapy improves the outcomes of metastatic colorectal cancer patients. Its most significant and dose-limiting side effect is the development of a neuropathic syndrome. The mechanism of the neurotoxicity is unclear. The limited knowledge about differences existing between neurotoxic and antitumor effects hinders the discovery of effective and safe adjuvant therapies. In vitro, we suggested cell-specific activation apoptotic pathways in normal nervous cells (astrocytes) vs. colon-cancer cells (HT-29). In the present research we compared the apoptotic signals evoked by oxaliplatin in astrocytes and HT-29 analyzing the intrinsic and extrinsic apoptotic pathways. In astrocytes, oxaliplatin induced a mitochondrial derangement measured as cytosolic release of cytochrome C, increase in superoxide anion levels and decreased expression of the antiapoptotic protein Bcl-2. Caspase-8, a main initiator of the extrinsic process remained unaltered. On the contrary, in HT-29 oxaliplatin increased caspase-8 activity and Bid expression, thus activating the extrinsic apoptosis, while the Bcl-2 increased expression blocked the mitochondrial damage. Data suggest the preferred activation of the intrinsic apoptosis as oxaliplatin damage signaling in normal nervous cells. The extrinsic pathway prevails in tumor cells indicating a possible strategy for planning new molecules to treat oxaliplatin-dependent neurotoxicity without negatively influence chemotherapy.
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Copper(II) sulfate solution, 4 % (w/v) (prepared from copper (II) sulfate pentahydrate)