Merck
  • Effects of subchronic exposure to glyphosate in juvenile oysters (Crassostrea gigas): From molecular to individual levels.
Effects of subchronic exposure to glyphosate in juvenile oysters (Crassostrea gigas): From molecular to individual levels.
Marine pollution bulletin (2014-12-03)
Antoine Mottier, Alexis Séguin, Alexandre Devos, Charles Le Pabic, Claire Voiseux, Jean Marc Lebel, Antoine Serpentini, Bruno Fievet, Katherine Costil
ABSTRACT
Glyphosate-based herbicides are extensively used and can be measured in aquatic ecosystems, including coastal waters. The effect of glyphosate on non-target organisms is an issue of worldwide concern. The aim of this study was to investigate the effects of subchronic exposure to glyphosate in juvenile oysters, Crassostrea gigas. Yearling oysters were exposed to three concentrations of glyphosate (0.1, 1 and 100μgL(-1)) for 56days. Various endpoints were studied, from the individual level (e.g., gametogenesis and tissue alterations) to the molecular level (mRNA quantification), including biochemical endpoints such as glutathione-S-transferase (GST) and catalase activities and malondialdehyde content. No mortality and growth occurred during the experiment, and individual biomarkers revealed only slight effects. The levels of gene expression significantly increased in oysters exposed to the highest glyphosate concentration (GST and metallothioneins) or to all concentrations (multi-xenobiotic resistance). These results suggested an activation of defence mechanisms at the molecular level.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
N-(Phosphonomethyl)glycine, 96%
Sigma-Aldrich
N-(Phosphonomethyl)glycine, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%