Merck
Evaluation of microbial quality and yeast diversity in fresh-cut apple.
Food microbiology (2015-07-19)
Ana Graça, David Santo, Eduardo Esteves, Carla Nunes, Maribel Abadias, Célia Quintas
ABSTRACT
The present work's aim was to study the microbial quality of minimally processed apples commercialized in Portugal. Sixty eight samples of fresh-cut apple were analyzed before their best-before date in 2011 and 2012 for aerobic mesophilic and psychrotrophic microorganisms, total coliforms, lactic-acid bacteria (LAB), coagulase-positive staphylococci and fungi. The parameters of food safety studied were Cronobacter sakazakii, Salmonella spp. and Listeria sp. Samples were analyzed according to standard methodologies and using Chromocult Agar for coliforms and Escherichia coli. The yeasts were identified by restriction analysis of the ITS-5.8S rDNA-region and 26S rDNA partial sequencing. The mesophilic and psychrotrophic microorganisms ranged from 3.3 to 8.9 and from 4.9 to 8.4 log CFU/g, respectively. Coliforms were detected in all the samples and staphylococci in 5.8% of them. LAB numbers varied from 2.8 to 8.7 and fungi (yeast and molds) from 3.6 to 7.1 log CFU/g. The most common yeasts were Candida sake and Pichia fermentans followed by Hanseniaspora spp., Candida spp., Meyerozyma guilliermondii, Metschnikowia pulcherrima, Cryptococcus spp. and the psychrotrophic Cystofilobasidium infirmominiatum. Foodborne bacteria and opportunistic pathogenic yeasts were not detected in the apples studied. The results obtained respected the European Commission regulation regarding criteria of food hygiene and safety.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis