Merck
  • Serum vitamin B12 levels during the first trimester of pregnancy correlate with newborn screening markers of vitamin B12 deficiency.
Serum vitamin B12 levels during the first trimester of pregnancy correlate with newborn screening markers of vitamin B12 deficiency.
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition (2014-01-01)
Anita Dayaldasani, Juan Ruiz-Escalera, Manuel Rodríguez-Espinosa, Inmaculada Rueda, Vidal Pérez-Valero, Raquel Yahyaoui
ABSTRACT
Low maternal vitamin B12 status is a risk factor for various adverse pregnancy outcomes. Although vitamin B12 deficiency is not a primary target of newborn screening (NBS) programs, measurements of propionylcarnitine (C3) and its ratios with acetylcarnitine (C3/C2) and palmitoylcarnitine (C3/C16) may incidentally identify vitamin B12-deficient newborns. The objective of this study was to measure vitamin B12 levels in women during the first trimester of pregnancy, evaluate predictors of these concentrations, and study their relationship with newborn screening results. Vitamin B12 concentrations were evaluated in 204 women during the first trimester of pregnancy and possible confounding factors were analyzed. After giving birth, data of their newborns (189) were collected (sex, gestational age, birthweight) and the acylcarnitine profile obtained by tandem mass spectrometry during NBS was analyzed. To assess the effects of the variables on vitamin B12 serum concentrations and newborn screening markers, stepwise multiple linear regression models were used. The mean serum concentration of vitamin B12 was 370.8 pmol/L (502.4 pg/mL) (SD 142.81). Vitamin B12 concentrations were significantly lower in smokers (p=0.027), and in women with low meat consumption (p=0.040). There was a significant inverse correlation between mothers'’ vitamin B12 concentrations and their children’'s C3 (r=-0.24; p=0.001), C3/C2 (r=-0.23; p=0.002) and C3/C16 levels (r=-0.20; p=0.006). Newborn screening markers (C3, C3/C2, and C3/C16) present an inverse correlation with maternal vitamin B12 status in the first trimester of pregnancy. Regarding factors that may influence maternal serum vitamin B12 levels during the first trimester, smoking seems to have a negative effect, and meat consumption a positive effect.
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%