Merck
  • Palmarosa [Cymbopogon martinii (Roxb.) Wats.] as a putative crop for phytoremediation, in tannery sludge polluted soil.
Palmarosa [Cymbopogon martinii (Roxb.) Wats.] as a putative crop for phytoremediation, in tannery sludge polluted soil.
Ecotoxicology and environmental safety (2015-08-25)
Janhvi Pandey, Sukhmal Chand, Shipra Pandey, Rajkumari, D D Patra
ABSTRACT
A field experiment using tannery sludge as a soil amendment material and palmarosa (Cymbopogon martinii) as a potential phytostabilizer was conducted to investigate their synergistic effect in relation to the improvement in soil quality/property. Three consecutive harvests of two cultivars of palmarosa-PRC-1 and Trishna, were examined to find out the influence of different tannery sludge doses on their herb, dry matter, essential oil yield and heavy metal accumulation. Soil fertility parameters (N, P, K, Organic carbon) were markedly affected by different doses of sludge. Enhanced soil nitrogen was positively correlated with herb yield (0.719*) and plant height (0.797*). The highest dose of tannery sludge (100 t ha(-1)) exhibited best performance than other treatments with respect to herb, dry matter and oil yield in all three harvests. Trishna was found to be superior to PRC-1 in relation to same studied traits. Quality of oil varied, but was insignificant statistically. Uptake of heavy metals followed same order (Cr>Ni>Pb>Cd) in roots and shoots. Translocation factor <1 for all trace elements and Bioconcentration factor >1 was observed in case of all heavy metals. Overall, tannery sludge enhanced the productivity of crop and metal accumulation occurred in roots with a meager translocation to shoots, hence it can be used as a phytostabiliser. The major advantage of taking palmarosa in metal polluted soil is that unlike food and agricultural crops, the product (essential oil) is extracted by hydro-distillation and there is no chance of oil contamination, thus is commercially acceptable.
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)