Merck
  • Hemodynamic shear stress modulates endothelial cell autophagy: Role of LOX-1.
Hemodynamic shear stress modulates endothelial cell autophagy: Role of LOX-1.
International journal of cardiology (2015-02-24)
Zufeng Ding, Shijie Liu, Xiaoyan Deng, Yubo Fan, Xianwei Wang, Jawahar L Mehta
ABSTRACT
Shear stress, autophagy and LOX-1 are important players in atherogenesis. Direct impact of shear stress on autophagy development in endothelial cells and role of LOX-1 therein are undelineated. A parallel-plate flow chamber was used to vary shear stress (3 to 30 dyn/cm(2)), and determine autophagy in endothelial cells. We observed that low shear stress (3 dyn/cm(2)) enhanced autophagy (expression of LC3-II) 2-3 fold, and increasing shear stress (15 to 30 dyn/cm(2)) resulted in a gradual decline. Autophagy increased when cells were treated with an inflammatory stimulus lipopolysaccharide (LPS). LOX-1 expression paralleled autophagy development. The in vitro observations were confirmed in the in vivo setting by studying autophagy (LC3-II and Beclin-1) and LOX-1 expression in wild-type mice given LPS. Expression of both autophagy and LOX-1 was most pronounced in aorto-iliac bifurcation region where shear stress is lower compared with aortic arch, thoracic aorta and iliac artery. To define the role of LOX-1 in the development of autophagy, we studied LOX-1 knockout mice. These mice despite LPS administration exhibited less autophagy (vs. wild-type mice). Role of LOX-1 in the regulation of autophagy was further established with LOX-1 inhibition (siRNA transfection and use of antibody) or overexpression (cDNA transfection), which showed that LOX-1 knockdown reduced while LOX-1 overexpression enhanced LC3-II expression in endothelial cells. These observations suggest that low shear stress is a powerful regulator on autophagy, particularly in state of inflammation, and LOX-1 plays an important role in shear stress induced autophagy.
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%
Sigma-Aldrich
4′-Hydroxy-3′-methoxyacetophenone, 98%
Sigma-Aldrich
Acetovanillone, ≥98%, FG
Sigma-Aldrich
MISSION® esiRNA, targeting human OLR1