Merck
  • Discriminating Lamiophlomis rotata According to Geographical Origin by (1)H-NMR Spectroscopy and Multivariate Analysis.
Discriminating Lamiophlomis rotata According to Geographical Origin by (1)H-NMR Spectroscopy and Multivariate Analysis.
Phytochemical analysis : PCA (2015-02-20)
Zheng Pan, Gang Fan, Rong-ping Yang, Wei-zao Luo, Xiang-dong Zhou, Yi Zhang
ABSTRACT
Lamiophlomis rotata (Duyiwei) is a folk herbal medicine that traditionally has been used in China as a hemostatic agent. Raw plant materials used for medicinal products from different geographical regions are often inconsistent in chemical composition. Metabolic fingerprinting provides a new approach for distinguishing the geographical origins of L. rotata. To identify metabolites that contribute to the different geographical regions of L. rotata samples. Lamiophlomis rotata metabolomics were performed by (1)H-nuclear magnetic resonance (NMR) spectroscopy and multivariate statistical analyses. The L. rotata metabolic profile was prepared for NMR measurements using methanol-d4 solvent. Principal component analysis (PCA) and hierarchical cluster analysis (HCA) were applied to analyse the L. rotata (1)H-NMR spectroscopy data. Nine iridoid glycosides, one flavonoid and three phenylpropanoid glycosides were detected in L. rotata by (1)H-NMR spectroscopy. (1)H-NMR measurements and multivariate analysis were used to successfully discriminate samples from three different locations. The NMR-based analysis of L. rotata is a more comprehensive approach than traditional chromatographic methods. Simple sample preparation, rapidity and reproducibility of are additional advantages of NMR analysis.
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, anhydrous, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, "100%", 99.96 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, 99 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS