Merck
  • Antitumor Activity of KW-2450 against Triple-Negative Breast Cancer by Inhibiting Aurora A and B Kinases.
Antitumor Activity of KW-2450 against Triple-Negative Breast Cancer by Inhibiting Aurora A and B Kinases.
Molecular cancer therapeutics (2015-10-08)
Kazuharu Kai, Kimie Kondo, Xiaoping Wang, Xuemei Xie, Mary K Pitner, Monica E Reyes, Angie M Torres-Adorno, Hiroko Masuda, Gabriel N Hortobagyi, Chandra Bartholomeusz, Hideyuki Saya, Debu Tripathy, Subrata Sen, Naoto T Ueno
ABSTRACT
Currently, no targeted drug is available for triple-negative breast cancer (TNBC), an aggressive breast cancer that does not express estrogen receptor, progesterone receptor, or HER2. TNBC has high mitotic activity, and, because Aurora A and B mitotic kinases drive cell division and are overexpressed in tumors with a high mitotic index, we hypothesized that inhibiting Aurora A and B produces a significant antitumor effect in TNBC. We tested this hypothesis by determining the antitumor effects of KW-2450, a multikinase inhibitor of both Aurora A and B kinases. We observed significant inhibitory activities of KW-2450 on cell viability, apoptosis, colony formation in agar, and mammosphere formation in TNBC cells. The growth of TNBC xenografts was significantly inhibited with KW-2450. In cell-cycle analysis, KW-2450 induced tetraploid accumulation followed by apoptosis or surviving octaploid (8N) cells, depending on dose. These phenotypes resembled those of Aurora B knockdown and complete pharmaceutical inhibition of Aurora A. We demonstrated that 8N cells resulting from KW-2450 treatment depended on the activation of mitogen-activated protein kinase kinase (MEK) for their survival. When treated with the MEK inhibitor selumetinib combined with KW-2450, compared with KW-2450 alone, the 8N cell population was significantly reduced and apoptosis was increased. Indeed, this combination showed synergistic antitumor effect in SUM149 TNBC xenografts. Collectively, Aurora A and B inhibition had a significant antitumor effect against TNBC, and this antitumor effect was maximized by the combination of selumetinib with Aurora A and B inhibition.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Aurkb
Sigma-Aldrich
MISSION® esiRNA, targeting human AURKB