MilliporeSigma
  • Quantification of soil/dust (SD) on the hands of children from Hubei Province, China using hand wipes.
Quantification of soil/dust (SD) on the hands of children from Hubei Province, China using hand wipes.
Ecotoxicology and environmental safety (2015-06-19)
Siyu Wang, Jin Ma, Libo Pan, Chunye Lin, Beibei Wang, Xiaoli Duan
ABSTRACT
A total of 120 children (58 males and 62 females) between the ages of 2 and 17 years were randomly selected from Wuhan City and Wufeng County in Hubei Province, China. We gathered hand SD samples from these children using hand wipes. We determined approximate amounts of hand SD and concentrations of three tracer soil elements (Ce, Y, V) in these samples. The approximate amounts of hand SD ranged from 6.35 to 85.42mg with a median value of 20.62mg. In addition, mean amounts of hand SD estimated using concentrations of Ce, Y, and V in samples were 1.07, 1.00, and 0.92mg, respectively. The amounts of hand SD varied greatly among age groups: primary school children had more hand SD than kindergarten and middle school children, males had more hand SD than females, and children from rural areas had more hand SD than those from urban areas. The rates of daily ingestion of hand SD for kindergarten, primary school, and middle school children were estimated as 1.79, 2.12, and 0.49mg/d, respectively.