Merck
  • Optimization of Surface Coating on Small Pd Nanosheets for in Vivo near-Infrared Photothermal Therapy of Tumor.
Optimization of Surface Coating on Small Pd Nanosheets for in Vivo near-Infrared Photothermal Therapy of Tumor.
ACS applied materials & interfaces (2015-06-16)
Saige Shi, Yizhuan Huang, Xiaolan Chen, Jian Weng, Nanfeng Zheng
ABSTRACT
Palladium nanosheets with strong near-infrared absorption have been recently demonstrated as promising photothermal agents for photothermal therapy (PTT) of cancers. However, systematic assessments of their potential risks and impacts to biological systems have not been fully explored yet. In this work, we carefully investigate how surface coatings affect the in vivo behaviors of small Pd nanosheets (Pd NSs). Several biocompatible molecules such as carboxymethyl chitosan (CMC), PEG-NH2, PEG-SH, and dihydrolipoic acid-zwitterion (DHLA-ZW) were used to coat Pd NSs. The blood circulation half-lives, biodistribution, potential toxicity, clearance, and photothermal effect of different surface-coated Pd NSs in mice after intravenous injection were compared. PEG-SH-coated Pd NSs (Pd-HS-PEG) were found to have ultralong blood circulation half-life and show high uptake in the tumor. We then carry out the in vivo photothermal therapeutic studies on the Pd-HS-PEG conjugate and revealed its outstanding efficacy in in vivo photothermal therapy of cancers. Our results highlight the importance of surface coatings to the in vivo behaviors of nanomaterials and can provide guidelines to the future design of Pd NSs bioconjugates for other in vivo applications.
Sigma-Aldrich
N,N-Dimethylpropionamide, 98%
Sigma-Aldrich
Dess-Martin periodinane, 97%