Merck
Isolation and characterization of mesenchymal stem cells.
Methods in molecular biology (Clifton, N.J.) (2014-01-30)
Sedat Odabas, A Eser Elçin, Y Murat Elçin
ABSTRACT
Mesenchymal stem cells (MSCs) have drawn great interest in the field of regenerative medicine, for cell replacement, immunomodulatory, and gene therapies. It has been shown that these multipotent stromal cells can be isolated from tissues such as bone marrow, adipose tissue, trimester amniotic tissue, umbilical cord blood, and deciduous teeth and can be expanded in adherent culture. They have the capacity to differentiate into cells of the connective tissue lineages in vitro and contribute to tissue parenchyma in vivo. However, proper in vitro manipulation of MSCs is a key issue to reveal a potential therapeutic benefit following transplantation into the patients. This chapter summarizes some of the essential protocols and assays used at our laboratory for the isolation, culture, differentiation, and characterization of mesenchymal stem cells from the bone marrow and adipose tissue.
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Bone Morphogenetic Protein 2 human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Transforming Growth Factor-β1 human, TGF-β1, recombinant, expressed in CHO cells, powder, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid tetrasodium salt dihydrate, BioReagent, suitable for cell culture, 98.5-102.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%