Merck
  • Functional polymorphism in ALOX15 results in increased allele-specific transcription in macrophages through binding of the transcription factor SPI1.
Functional polymorphism in ALOX15 results in increased allele-specific transcription in macrophages through binding of the transcription factor SPI1.
Human mutation (2005-12-02)
Jonas Wittwer, Jacqueline Marti-Jaun, Martin Hersberger
ABSTRACT
The reticulocyte-type 15-lipoxygenase-1 (ALOX15) has antiinflammatory and inflammatory effects, and is implicated in the development of asthma, arthritis, and atherosclerosis. We screened the human ALOX15 gene for variations because genetic variability in ALOX15 may influence these diseases. We detected 11 variations, including five polymorphisms located in the ALOX15 promoter region. One of these polymorphisms, a C-to-T substitution at position c.-292, created a novel transcription factor binding site for SPI1. Transcription assays revealed that promoter variants with c.-292 T transcribe twice as efficiently as all the other promoter variants containing c.-292C. This was true in macrophages that constitutively express SPI1, but not in a lung epithelial cell line that does not express SPI1. Mutation of the core-binding site for SPI1 abolished the higher transcriptional activity, and electrophoretic mobility shift assays showed that SPI1 selectively binds to the mutant c.-292 T [corrected] promoter. These results were corroborated in primary human macrophages, in which macrophages from heterozygous c.-292CT carriers expressed three times more ALOX15 mRNA than macrophages from homozygous c.-292CC carriers. We conclude that the c.-292 T allele in the ALOX15 promoter generates a novel binding site for the transcription factor SPI1 that results in higher transcription of the gene in macrophages. This may lead to an increase in ALOX15-mediated lipid metabolites, which play a role in inflammation.