Merck
Gap junction proteins in the light-damaged albino rat.
Molecular vision (2014-06-03)
Cindy X Guo, Henry Tran, Colin R Green, Helen V Danesh-Meyer, Monica L Acosta
ABSTRACT
Changes in connexin expression are associated with many pathological conditions seen in animal models and in humans. We hypothesized that gap junctions are important mediators in tissue dysfunction and injury processes in the retina, and therefore, we investigated the pattern of connexin protein expression in the light-damaged albino rat eye. Adult Sprague-Dawley rats were exposed to intense light for 24 h. The animals were euthanized, and ocular tissue was harvested at 0 h, 6 h, 24 h, 48 h, and 7 days after light damage. The tissues were processed for immunohistochemistry and western blotting to analyze the expression of the gap junction proteins in the light-damaged condition compared to the non-light-damaged condition. Cell death was detected using the terminal deoxynucleotidyl transferase-mediated dUTP nick-end labeling (TUNEL) technique. Intense light exposure caused increased TUNEL labeling of photoreceptor cells. Immunocytochemistry revealed that connexin 36 (Cx36) was significantly increased in the inner plexiform layer and Cx45 was significantly decreased in the light-damaged retina. The pattern of Cx36 and Cx45 labeling returned to normal 7 days after light damage. Cx43 significantly increased in the RPE and the choroid in the light-damaged tissue, and decreased but not significantly in the retina. This elevated Cx43 expression in the choroid colocalized with markers of nitration-related oxidative stress (nitrotyrosine) and inflammation (CD45 and ionized calcium-binding adaptor molecule-1) in the choroid. The results suggest that connexins are regulated differently in the retina than in the choroid in response to photoreceptor damage. Changes in connexins, including Cx36, Cx43, and Cx45, may contribute to the damage process. Specifically, Cx43 was associated with inflammatory damage. Therefore, connexins may be candidate targets for treatment for ameliorating disease progression.
Sigma-Aldrich
Monoclonal Anti-GJA1 antibody produced in mouse, clone 3E5, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Nitrotyrosine Antibody, clone 1A6, clone 1A6, Upstate®, from mouse
Sigma-Aldrich
Anti-Connexin-43 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
DAPI, for nucleic acid staining
Fluorescein, European Pharmacopoeia (EP) Reference Standard