Merck
  • A robust synthesis of N-glycolyl muramyl dipeptide via azidonitration/reduction.
A robust synthesis of N-glycolyl muramyl dipeptide via azidonitration/reduction.
Organic & biomolecular chemistry (2014-12-06)
Shuo Xing, James L Gleason
ABSTRACT
A novel synthetic route leading to N-glycolyl muramyl dipeptide (MDP), a bacterial glycopeptide of particular interest in studies of nucleotide-binding oligomerization domain-containing protein 2 (NOD2), is described. The synthetic strategy hinges on the alkylation of benzylidene-protected glucal with 2-bromopropionic acid and thus circumvents a challenging and non-reproducible SN2 step at the C-3 position of glucosamine derivatives. The subsequent sequence includes an azidonitration and an unusual azide reduction/acylation step via an aza ylide/oxaphospholidine intermediate. This approach generates a protected N-glycolyl MDP that can be either subjected to a one-step global deprotection or differentially deprotected to obtain further derivatives.
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Sodium hydride, dry, 90%