Merck
In vitro antiplasmodial activity of cepharanthine.
Malaria journal (2014-08-26)
Camille Desgrouas, Charles Chapus, Jérôme Desplans, Christelle Travaille, Aurélie Pascual, Béatrice Baghdikian, Evelyne Ollivier, Daniel Parzy, Nicolas Taudon
ABSTRACT
New classes of anti-malarial drugs are needed to control the alarming Plasmodium falciparum resistance toward current anti-malarial therapy. The ethnopharmacological approach allows the discovery of original chemical structures from the vegetable biodiversity. Previous studies led to the selection of a bisbenzylisoquinoline, called cepharanthine and isolated from a Cambodian plant: Stephania rotunda. Cepharanthine could exert a mechanism of action different from commonly used drugs. Potential plasmodial targets are reported here. To study the mechanism of action of cepharanthine, a combined approach using phenotypic and transcriptomic techniques was undertaken. Cepharanthine blocked P. falciparum development in ring stage. On a culture of synchronized ring stage, the comparisons of expression profiles showed that the samples treated with 5 μM of cepharanthine (IC90) were significantly closer to the initial controls than to the final ones. After a two-way ANOVA (p-value < 0.05) on the microarray results, 1,141 probes among 9,722 presented a significant differential expression.A gene ontology analysis showed that the Maurer's clefts seem particularly down-regulated by cepharanthine. The analysis of metabolic pathways showed an impact on cell-cell interactions (cytoadherence and rosetting), glycolysis and isoprenoid pathways. Organellar functions, more particularly constituted by apicoplast and mitochondrion, are targeted too. The blockage at the ring stage by cepharanthine is described for the first time. Transcriptomic approach confirmed that cepharanthine might have a potential innovative antiplasmodial mechanism of action. Thus, cepharanthine might play an ongoing role in the progress on anti-malarial drug discovery efforts.
Product Description
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Pricing and availability is not currently available.
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-Sorbitol, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Pricing and availability is not currently available.