Merck
  • Acetonitrile adduct formation as a sensitive means for simple alcohol detection by LC-MS.
Acetonitrile adduct formation as a sensitive means for simple alcohol detection by LC-MS.
Journal of the American Society for Mass Spectrometry (2014-09-25)
Roy Bogseth, Eric Edgcomb, Christopher M Jones, Edward K Chess, Peifeng Hu
ABSTRACT
Simple alcohols formed protonated acetonitrile adducts containing up to two acetonitrile molecules when analyzed by ESI or APCI in the presence of acetonitrile in the solvent. These acetonitrile adducts underwent dissociation to form a nitrilium ion, also referred to as the substitution ion. Diols and triols behaved differently. In ESI, they formed only one acetonitrile adduct containing one acetonitrile. The S ion was not observed in ESI and was only weakly observed from the dissociation of the (M + ACN + H)(+) ion. On the other hand, the S ion was abundantly formed from the diols in APCI. This formation of acetonitrile adducts and substitution ion from simple alcohols/diols offers an opportunity to detect simple alcohols/diols sensitively by LC-MS interfaced by ESI or APCI. The utility of this chemistry was demonstrated in a method developed for the quantification of cyclohexanol in rat plasma by monitoring the CID-induced fragmentation from the S ion to a fragment ion.
Product Description
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,5-Pentanediol, 96%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Supelco
1-Propanol, analytical standard
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
1,5-Pentanediol, purum, ≥97.0% (GC)
Supelco
1-Octanol, analytical standard
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
1-Octanol, suitable for HPLC, ≥99%
Supelco
1-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
USP
1-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%