Merck
  • Protein disulphide-isomerase A2 regulated intracellular tissue factor mobilisation in migrating human vascular smooth muscle cells.
Protein disulphide-isomerase A2 regulated intracellular tissue factor mobilisation in migrating human vascular smooth muscle cells.
Thrombosis and haemostasis (2015-01-30)
Esther Peña, Gemma Arderiu, Lina Badimon
ABSTRACT
Protein-disulphide isomerase family (PDI) are an ER-stress protein that controls TF-procoagulant activity but its role in HVSMC migration and coronary artery disease remains to be elucidated. We aimed to investigate whether in human coronary smooth muscle cells (HVSMC) the ER-stress protein-disulphide isomerase family A member 2 (PDIA2) regulates tissue factor (TF) polarisation during migration and atherosclerotic remodeling. PDIA2 and TF were analysed by confocal microscopy, silenced by small interfering RNAs (siRNA) and their function analysed by transwell and migration assays in vitro and in vivo. PDIA2and TF co-localise in the front edge of motile HVSMC. Silencing PDIA2, as well as silencing TF, reduces migration. PDIA2 silenced cells show increased TF-rich microparticle shedding. In vivo cell-loaded plug implants in nude mice of PDIA2 silenced HVSMC together with microvascular endothelial cells showed a significant impairment in mature microvessel formation. PDIA2 and TF are found in remodelled atherosclerotic plaques but not in healthy coronaries. In conclusion, we demonstrate that TF is chaperoned by PDIA2 to the HVSMC membrane and to the cell migratory front. Absence of PDIA2 impairs TF intracellular trafficking to its membrane docking favoring its uncontrolled release in microparticles. TF-regulated HVSMC migration and microvessel formation is under the control of the ER-protein PDIA2.
Sigma-Aldrich
MISSION® esiRNA, targeting human PDIA2 (2)
Sigma-Aldrich
MISSION® esiRNA, targeting human PDIA2 (1)
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
MISSION® esiRNA, targeting human TF
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pdia2
Sigma-Aldrich
MISSION® esiRNA, targeting human F3