Merck
  • Enantioselective synthesis of benzoindolizidine derivatives using chiral phase-transfer catalytic intramolecular domino aza-Michael addition/alkylation.
Enantioselective synthesis of benzoindolizidine derivatives using chiral phase-transfer catalytic intramolecular domino aza-Michael addition/alkylation.
Organic & biomolecular chemistry (2014-11-28)
Jiajia Guo, Shouyun Yu
ABSTRACT
An efficient and enantioselective strategy to synthesize benzoindolizidines from α,β-unsaturated amino ketones via domino intramolecular aza-Michael addition/alkylation was developed. These reactions were enabled by cinchona alkaloid-derived quaternary ammonium salts as the phase-transfer catalyst. A variety of benzoindolizidines were prepared in good yields (up to 93%) and enantioselectivities (up to 92.8:7.2 er).
Sigma-Aldrich
3-Hydroxyisoquinoline, 99%