Merck
  • Effects of eye drops containing a mixture of omega-3 essential fatty acids and hyaluronic acid on the ocular surface in desiccating stress-induced murine dry eye.
Effects of eye drops containing a mixture of omega-3 essential fatty acids and hyaluronic acid on the ocular surface in desiccating stress-induced murine dry eye.
Current eye research (2014-02-25)
Zhengri Li, Jung-Han Choi, Han-Jin Oh, Soo-Hyun Park, Jee-Bum Lee, Kyung Chul Yoon
ABSTRACT
To investigate the efficacy of the topical application of omega-3 essential fatty acids (EFAs) and hyaluronic acid (HA) mixtures in a mouse model of experimental dry eye (EDE). Eye drops consisting of 0.1% HA, 0.02%, or 0.2% omega-3 EFAs alone and mixture of 0.02%, or 0.2% omega-3 EFAs and 0.1% HA were applied in desiccating stress-induced murine dry eye. Corneal irregularity scores and fluorescein staining scores were measured 5 and 10 days after treatment. Levels of interleukin (IL)-1β, -17, and interferon gamma-induced protein (IP)-10 were measured in the conjunctiva at 10 days using a multiplex immunobead assay. The concentrations of hexanoyl-lys (HEL) and 4-hydroxynonenal (4-HNE) in conjunctiva tissue were measured with enzyme-linked immunosorbent assays. Mice treated with the mixture containing 0.2% omega-3 EFAs showed a significant improvement in corneal irregularity scores and corneal fluorescein staining scores compared with EDE, HA, 0.02% or 0.2% omega-3 EFAs alone, and 0.02% omega-3 EFAs mixture-treated mice. A significant decrease in the levels of IL-1β, -17, and IP-10 were observed in the 0.2% EFAs mixture-treated group, compared with the other groups. In the mice treated with the mixture containing 0.2% omega-3 EFAs, the concentration of 4-HNE was also lower than the other groups. Although 0.2% omega-3 EFAs alone group also had a significant improvement in corneal irregularity scores and IL-17, IL-10, and 4 HNE levels compared with the other groups, the efficacy was lower than 0.2% omega-3 mixture group. Topically applied eye drops containing a mixture of omega-3 EFAs and HA could improve corneal irregularity and corneal epithelial barrier disruption, and decrease inflammatory cytokines and oxidative stress markers on the ocular surface. Topical omega-3 EFAs and HA mixture may have a greater therapeutic effect on clinical signs and inflammation of dry eye compared with HA artificial tears.
Sigma-Aldrich
(−)-Scopolamine hydrobromide trihydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Scopolamine hydrobromide, meets USP testing specifications