Merck
  • Ex-vivo permeation study of chlorin e6-polyvinylpyrrolidone complexes through the chick chorioallantoic membrane model.
Ex-vivo permeation study of chlorin e6-polyvinylpyrrolidone complexes through the chick chorioallantoic membrane model.
The Journal of pharmacy and pharmacology (2014-04-05)
Romchat Chutoprapat, Lai W Chan, Paul W S Heng
ABSTRACT
To investigate the influence of the hydrophilic polymer, polyvinylpyrrolidone (PVP) on the ex-vivo permeability of the poorly water-soluble photosensitizer, chlorin e6 (Ce6) using the chick chorioallantoic membrane (CAM) model. The CAM was removed from the fertilized chicken egg at embryo age of 15 days. The permeation profiles of Ce6 and PVP complexes (Ce6-PVP) at 1:0, 1:1, 1:10, 1:50 and 1:100 w/w in different pH conditions were first studied using the CAM model with Franz diffusion cell over 8 h. The solution viscosity of the formulations and apparent solubility of Ce6 were also investigated. The permeability of Ce6 was found to be directly proportional to the amount of PVP used and the apparent solubility of Ce6. Permeability was only marginally affected by the solution viscosity of the formulations. The permeability of Ce6 was lowered in the acidic pH. Ce6-PVP at 1:100 w/w gave the highest percentage release of Ce6 across the CAM, with 23% at pH 3 and 55% at pH 7.4, after 8 h, respectively. The present work suggests that PVP had served as penetration enhancer for the poorly water-soluble Ce6 and the CAM can serve as a useful biological membrane model for preclinical permeability study of biological and pharmaceutical substances. The Ce6-PVP formulation at 1:100 w/w can be applied for the further clinical investigation.
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%