Merck
  • Effects of administering sodium selenite, methylseleninic acid, and seleno-L-methionine on glucose tolerance in a streptozotocin/nicotinamide-induced diabetic mouse model.
Effects of administering sodium selenite, methylseleninic acid, and seleno-L-methionine on glucose tolerance in a streptozotocin/nicotinamide-induced diabetic mouse model.
Biological & pharmaceutical bulletin (2014-09-02)
Hitoshi Ueno, Ryo Shimizu, Tomofumi Okuno, Hirofumi Ogino, Tomohiro Arakawa, Fumitoshi Sakazaki, Katsuhiko Nakamuro
ABSTRACT
The effects of administering the selenocompounds, sodium selenite, methylseleninic acid (MSA), and seleno-L-methionine (SeMet) on glucose tolerance were compared in the nicotinamide (NA) and streptozotocin (STZ)-induced diabetic mouse model. ICR mice were intraperitoneally treated twice with STZ (100 mg/kg) 15 min after an injection of NA (120 mg/kg) at a 1-d interval. Non-fasting blood glucose levels were then monitored weekly while orally administering the selenocompounds at 158 µg Se/kg body weight with free access to a selenium-deficient diet for 5 weeks. The mean body weights of NA/STZ-induced diabetic mice were partly restored by the administration of selenocompounds, while SeMet led to a higher selenium content and glutathione peroxidase 1 activity in the pancreas. Non-fasting and oral glucose tolerance-tested blood glucose levels, which were elevated by NA/STZ, were significantly suppressed by the administration of SeMet. These results suggest that SeMet may improve glucose tolerance in a NA/STZ-induced mild diabetic mouse model by increasing bioavailability in the pancreas.
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)