Merck
  • Dense fibrillar collagen is a potent inducer of invadopodia via a specific signaling network.
Dense fibrillar collagen is a potent inducer of invadopodia via a specific signaling network.
The Journal of cell biology (2015-02-04)
Vira V Artym, Stephen Swatkoski, Kazue Matsumoto, Catherine B Campbell, Ryan J Petrie, Emilios K Dimitriadis, Xin Li, Susette C Mueller, Thomas H Bugge, Marjan Gucek, Kenneth M Yamada
ABSTRACT
Cell interactions with the extracellular matrix (ECM) can regulate multiple cellular activities and the matrix itself in dynamic, bidirectional processes. One such process is local proteolytic modification of the ECM. Invadopodia of tumor cells are actin-rich proteolytic protrusions that locally degrade matrix molecules and mediate invasion. We report that a novel high-density fibrillar collagen (HDFC) matrix is a potent inducer of invadopodia, both in carcinoma cell lines and in primary human fibroblasts. In carcinoma cells, HDFC matrix induced formation of invadopodia via a specific integrin signaling pathway that did not require growth factors or even altered gene and protein expression. In contrast, phosphoproteomics identified major changes in a complex phosphosignaling network with kindlin2 serine phosphorylation as a key regulatory element. This kindlin2-dependent signal transduction network was required for efficient induction of invadopodia on dense fibrillar collagen and for local degradation of collagen. This novel phosphosignaling mechanism regulates cell surface invadopodia via kindlin2 for local proteolytic remodeling of the ECM.
Sigma-Aldrich
Anti-Integrin β2 Antibody, clone P4H9, azide free, clone P4H9, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α3 Antibody, clone P1B5, azide free, clone P1B5, Chemicon®, from mouse
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Anti-ARHGEF18 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ARHGEF12 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-TIAM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Monoclonal Anti-Collagen, Type I antibody produced in mouse, clone COL-1, ascites fluid
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Eosin Y disodium salt, Dye content ≥85 %
Sigma-Aldrich
Eosin Y disodium salt, certified by the Biological Stain Commission
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone P1E6, azide free, clone P1E6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Tiam1 Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Anti-Integrin α1 Antibody, clone FB12, azide free, clone FB12, Chemicon®, from mouse
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)