Merck
Silk fibroin rods for sustained delivery of breast cancer therapeutics.
Biomaterials (2014-07-11)
Tuna Yucel, Michael L Lovett, Raffaella Giangregorio, Erin Coonahan, David L Kaplan
ABSTRACT
A silk-protein based reservoir rod was developed for zero-order and long-term sustained drug delivery applications. Silk reservoir rod formulations were processed in three steps. First, a regenerated silk fibroin solution, rich in random-coil content was transformed into a tubular silk film with controllable dimensions, uniform film morphology and a structure rich in silk II, β-sheet content via "film-spinning." Second, the drug powder was loaded into swollen silk tubes followed by tube end clamping. Last, clamped silk tube ends were sealed completely via dip coating. Anastrozole, an FDA approved active ingredient for the treatment of breast cancer, was used as a model drug to investigate viability of the silk reservoir rod technology for sustained drug delivery. The in vitro and in vivo pharmacokinetic data (in a female Sprague-Dawley rat model) analyzed via liquid chromatography-tandem mass spectroscopy indicated zero-order release for 91 days. Both in vitro and in vivo anastrozole release rates could be controlled simply by varying silk rod dimensions. The swelling behavior of silk films and zero-order anastrozole release kinetics indicated practically immediate film hydration and formation of a linear anastrozole concentration gradient along the silk film thickness. The dependence of anastrozole release rate on the overall silk rod dimensions was in good agreement with an essentially diffusion-controlled sustained release from a reservoir cylindrical geometry. In vivo results highlighted a strong in vitro-in vivo pharmacokinetic correlation and a desirable biocompatibility profile of silk reservoir rods. During a 6-month implantation in rats, the apparent silk molecular weight values decreased gradually, while rod dry mass and β-sheet crystal content values remained essentially constant, providing a suitable timeframe for controlled, long-term sustained delivery applications. Overall, the silk reservoir rod may be a viable candidate for sustained delivery of breast cancer therapeutics.
Sigma-Aldrich
Anastrozole, ≥98% (HPLC)
Pricing and availability is not currently available.
Anastrozole, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.