Merck
  • Inhibition of opioid receptor mediated G-protein activity after chronic administration of kynurenic acid and its derivative without direct binding to opioid receptors.
Inhibition of opioid receptor mediated G-protein activity after chronic administration of kynurenic acid and its derivative without direct binding to opioid receptors.
CNS & neurological disorders drug targets (2014-12-06)
Ferenc Zador, Reza Samavati, Eszter Szlavicz, Bernadett Tuka, Engin Bojnik, Ferenc Fulop, Jozsef Toldi, Laszlo Vecsei, Anna Borsodi
ABSTRACT
There is an increasing number of evidence showing analgesic properties of the kynurenic acid (KYNA), and also some studies demonstrate that kynurenine might interact with the opioid system. Therefore in this study, for the first time we investigated the direct binding affinity of KYNA and its structural analog KYNA-1 towards mu, kappa and delta opioid receptor in competition binding experiments applying opioid receptor specific radioligands. The binding affinity measurements were performed in Chinese hamster ovary cell lines overexpressing the corresponding opioid receptor (mu and kappa opioid receptor were rat, delta opioid receptor were mouse sequence). Additionally we also examined the chronic effect of these compounds on mu, kappa and delta opioid receptor and also nociceptin peptide receptor mediated G-protein activity in [(35)S]GTPγS binding assays performed in mouse cortex and striatum membranes. Our results showed that KYNA and KYNA-1 had no affinity towards any of the three classic opioid receptors. On the other hand the compounds significantly decreased opioid and nociceptin receptor mediated G-protein activity or in some cases enhanced the potency of the activating ligand. Moreover, the alterations were receptor and brain region specific. Accordingly, we conclude that KYNA and KYNA-1 do not interact directly with the opioid receptors, but more likely alter the receptor functions intracellularly.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Kynurenic acid, ≥98%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl