Merck
  • A novel label-free electrochemiluminescence aptasensor based on layered flowerlike molybdenum sulfide-graphene nanocomposites as matrix.
A novel label-free electrochemiluminescence aptasensor based on layered flowerlike molybdenum sulfide-graphene nanocomposites as matrix.
Colloids and surfaces. B, Biointerfaces (2014-07-30)
Yan-Ming Liu, Gui-Fang Shi, Jing-Jing Zhang, Min Zhou, Jun-Tao Cao, Ke-Jing Huang, Shu-Wei Ren
ABSTRACT
A label-free and ultrasensitive electrochemiluminescence (ECL) aptasensor was constructed for the detection of thrombin. Molybdenum sulfide-graphene nanocomposites with good conductivity and large surface area were immobilized on glassy carbon electrode (GCE), and then Nafion was fixed to chemosorb the Ru(bpy)3(2+) used as luminescence agent. Subsequently, gold nanoparticles (AuNPs) were modified on the electrode to immobilize the thiol-modified thrombin aptamer for fabrication of the thrombin aptasensor. The proposed ECL aptasensor produced the ultrasensitive detection of thrombin with a low detection limit of 3.6×10(-15)M (S/N=3) and over a wide target concentration range from 1.0×10(-14) to 5.0×10(-9)M. The aptasensor has been successfully applied in the determination of thrombin in human plasma samples of both traumatic and non-traumatic injury patients, indicating its promise in biochemical analysis. The recoveries of thrombin in human plasma samples are between 88.6% and 105.0%, and the RSD values are no more than 3.7%. The results demonstrate that this aptasensor has excellent sensitivity, selectivity and stability.
Sigma-Aldrich
Trisodium citrate dihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium citrate tribasic dihydrate, puriss. p.a., ACS reagent, ≥99.0% (NT)
Sigma-Aldrich
Sodium citrate tribasic dihydrate, BioUltra, for molecular biology
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG
SAFC
L-Cysteine
Sigma-Aldrich
Sodium citrate tribasic dihydrate, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ≥98%
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%