Merck
  • A simple guide to the terminology and application of leucocyte monoclonal antibodies.
A simple guide to the terminology and application of leucocyte monoclonal antibodies.
Histopathology (1988-05-01)
J K Chan, C S Ng, P K Hui
ABSTRACT
This review aims to provide a simple guide and quick reference to the terminology and diagnostic applications of leucocyte monoclonal antibodies. The differentiation cluster terminology, where applicable, is used throughout.
Sigma-Aldrich
CD43 (MT1) Mouse Monoclonal Antibody