Merck
  • Cyclooxygenase-2-dependent oxidative stress mediates palmitate-induced impairment of endothelium-dependent relaxations in mouse arteries.
Cyclooxygenase-2-dependent oxidative stress mediates palmitate-induced impairment of endothelium-dependent relaxations in mouse arteries.
Biochemical pharmacology (2014-08-26)
Zhen Gao, Huina Zhang, Jian Liu, Chi Wai Lau, Pingsheng Liu, Zhen Yu Chen, Hung Kay Lee, George L Tipoe, Hing Man Ho, Xiaoqiang Yao, Yu Huang
ABSTRACT
Palmitic acid, one of the saturated free fatty acids, impairs cardiovascular function as manifested by inducing vascular inflammation, apoptosis and over-production of reactive oxygen species (ROS) although the origin for ROS remains unclear. The present study investigated the cellular mechanisms underlying palmitate-induced impairment of endothelial function. Ex vivo treatment in tissue culture with palmitate concentration-dependently attenuated acetylcholine-induced endothelium-dependent relaxations, up-regulated the expression of cyclooxygenase-2 (COX-2) and elevated superoxide formation in mouse aortic endothelial cells (MAECs) measured by dihydroethidium (DHE) staining and electron paramagnetic resonance (EPR) spectroscopy. Superoxide scavengers, COX-2 inhibitor and thromboxane prostanoid (TP) receptor antagonist, but not COX-1 inhibitor reversed the harmful effects of palmitate. Furthermore, palmitate impaired acetylcholine-induced relaxations and raised superoxide in en face endothelium of aortas only from COX-1(-/-) mice but not from COX-2(-/-) mice. Palmitate increased the production and release of TXB2, a stable thromboxane A2 metabolite in mouse aortas, which was abolished by COX-2 inhibitor. Superoxide scavenger did not affect palmitate-induced up-regulated expression of COX-2 in MAECs. Both real time PCR and luciferase reporter gene assay confirmed COX-2 up-regulation in palmitate-treated MAECs and NF-κB was substantially involved in this up-regulation. The present study provides novel evidence that palmitate up-regulates COX-2 through NF-κB-dependent mechanism and resultant COX-2-associated oxidative stress impairs endothelium-dependent relaxations in mouse aortas.
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, for complexometry, ≥99.0%
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Palmitic acid, ≥99%
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Celecoxib, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium palmitate, ≥98.5%
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
Parthenolide, ≥98% (HPLC)
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Supelco
Sodium Nitroprusside, Pharmaceutical Secondary Standard; Certified Reference Material