Merck
  • Involvement of CaM-CaMKII-ERK in bisphenol A-induced Sertoli cell apoptosis.
Involvement of CaM-CaMKII-ERK in bisphenol A-induced Sertoli cell apoptosis.
Toxicology (2014-06-07)
Wenyi Qian, Jingying Zhu, Changfei Mao, Jingli Liu, Yixin Wang, Qiang Wang, Yanqing Liu, Rong Gao, Hang Xiao, Jun Wang
ABSTRACT
Bisphenol A (BPA), one of the most prevalent chemicals for daily use, has been reported as a xenoestrogen to induce reproductive toxicity, but its mechanism is poorly understood. In the present study, we aimed to explore whether CaM-CaMKII-ERK1/2 signaling pathway was involved in BPA-induced Sertoli cells injury via the mitochondrial apoptotic pathway. TM4 cells were cultured with 0, 0.02, 0.2, 2.0, 20μM BPA, and cell viability, mitochondrial function and CaM-CaMKII-ERK1/2 signal pathway were examined. With the MTT assay, BPA was found to suppress cell viability in a dose- and time-dependent manner. Moreover, mitochondrial mass loss, membrane potential decrease, cytochrome c release, Bcl-2 family members down-regulation and caspases-3 up-regulation were obviously observed when the TM4 cells were exposed to BPA. Additionally, the expression of calmodulin (CaM) and phosphorylation of calcium/calmodulin dependent kinase II (CaMKII) significantly increased, and pretreatment with 10μM antagonist of CaM (W-7) or CaMKII (KN62) prevented cell damage through mitochondrial apoptotic pathway. In parallel, ERK1/2 pathway was proved to participate in BPA-induced cell damage, since W-7 and KN62 partially suppressed ERK1/2 activation, and PD98059, the ERK1/2 antagonist, significantly attenuated BPA-induced cell damage. These data, taken together, indicated that CaM-CaMKII-ERK axis might transmit apoptotic signals to the mitochondria during BPA-induced cell apoptosis. By exploring the mechanisms of the Ca(2+) homeostasis and the corresponding proteins, our study provides new insight into BPA-induced reproductive toxicity.
Sigma-Aldrich
Cytochrome c from equine heart, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis
Sigma-Aldrich
Cytochrome c from pigeon breast muscle, ≥95% based on Mol. Wt. 12,173 basis
Sigma-Aldrich
Cytochrome c from equine heart, BioReagent, suitable for GFC marker
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% based on Mol. Wt. 12,384 basis
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Bisphenol A, 97%
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrome c from Saccharomyces cerevisiae, ≥85% based on Mol. Wt. 12,588 basis
Supelco
Digoxigenin, analytical standard
Sigma-Aldrich
ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation
Digoxigenin, European Pharmacopoeia (EP) Reference Standard