Merck
  • Azido- or hydroxyl-capped half-cubanes containing Cp*Rh fragments: [Cp*3Rh3(mu-X)3(mu3-X)]2+ (X-=OH- or N3-).
Azido- or hydroxyl-capped half-cubanes containing Cp*Rh fragments: [Cp*3Rh3(mu-X)3(mu3-X)]2+ (X-=OH- or N3-).
Dalton transactions (Cambridge, England : 2003) (2004-10-16)
Won Seok Han, Soon W Lee
ABSTRACT
Treatment of [Cp*Rh(H(2)O)(3)](OTf)(2) (1) with Me(3)SiNH-t-Bu in acetone gave a hydroxyl-capped half-cubane [Cp*(3)Rh(3)(mu-OH)(3)(mu(3)-OH)](OTf)(3)(t-BuNH(3)) (2). Slow diffusion of Me(3)SiN(3) in diethyl ether into compound in acetone produced an azido-capped half-cubane [Cp*(3)Rh(3)(mu-N(3))(3)(mu(3)-N(3))](OTf)(2) (3). On the other hand, treating 1 with Me(3)SiN(3) in acetone gave an azido-bridged, dinuclear rhodium(III) complex [Cp*Rh(mu-N(3))(OH(2))](2)(OTf)(2) (4). Complexes 2 and 3 represent the first azido- or hydroxyl-capped, incomplete cubane-type Rh clusters. Under appropriate conditions, complexes 2 and 3 could be converted to complex 4. The structures of all products were determined by X-ray diffraction.
Sigma-Aldrich
N-tert-Butyltrimethylsilylamine, 98%