Merck
  • Mid term results with the curved modular tapered, fluted titanium Revitan stem in revision hip replacement.
Mid term results with the curved modular tapered, fluted titanium Revitan stem in revision hip replacement.
The bone & joint journal (2014-07-06)
B Fink, K Urbansky, P Schuster
ABSTRACT
We report our experience of revision total hip replacement (THR) using the Revitan curved modular titanium fluted revision stem in patients with a full spectrum of proximal femoral defects. A total of 112 patients (116 revisions) with a mean age of 73.4 years (39 to 90) were included in the study. The mean follow-up was 7.5 years (5.3 to 9.1). A total of 12 patients (12 hips) died but their data were included in the survival analysis, and four patients (4 hips) were lost to follow-up. The clinical outcome, proximal bone regeneration and subsidence were assessed for 101 hips. The mean Harris Hip Score was 88.2 (45.8 to 100) after five years and there was an increase of the mean Barnett and Nordin-Score, a measure of the proximal bone regeneration, of 20.8 (-3.1 to 52.7). Five stems had to be revised (4.3%), three (2.9%) showed subsidence, five (4.3%) a dislocation and two of 85 aseptic revisions (2.3%) a periprosthetic infection. At the latest follow-up, the survival with revision of the stem as the endpoint was 95.7% (95% confidence interval 91.9% to 99.4%) and with aseptic loosening as the endpoint, was 100%. Peri-prosthetic fractures were not observed. We report excellent results with respect to subsidence, the risk of fracture, and loosening after femoral revision using a modular curved revision stem with distal cone-in-cone fixation. A successful outcome depends on careful pre-operative planning and the use of a transfemoral approach when the anatomy is distorted or a fracture is imminent, or residual cement or a partially-secured existing stem cannot be removed. The shortest appropriate stem should, in our opinion, be used and secured with > 3 cm fixation at the femoral isthmus, and distal interlocking screws should be used for additional stability when this goal cannot be realised.
Product Description
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Titanium, rod, 200mm, diameter 2mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 16mm, as drawn, 99.99+%
Titanium, microfoil, disks, 10mm, thinness 1.0μm, specific density 429μg/cm2, permanent mylar 3.5μm support, 99.6+%
Titanium, rod, 490mm, diameter 2mm, annealed, 99.6+%
Titanium, tube, 1220mm, outside diameter 2.03mm, inside diameter 1.55mm, wall thickness 0.24mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 25mm, diameter 16mm, as drawn, 99.99+%
Titanium, rod, 500mm, diameter 1.5mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 6mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 25.4mm, inside diameter 23.62mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 5mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 1.6mm, inside diameter 1.2mm, wall thickness 0.2mm, hard, 99.6+%
Titanium, rod, 200mm, diameter 4mm, annealed, 99.6+%
Titanium, rod, 1000mm, diameter 1.5mm, annealed, 99.6+%
Titanium, microfoil, disks, 25mm, thinness 0.25μm, specific density 112.6μg/cm2, permanent mylar 3.5μm support, 99.6+%
Titanium, tube, 100mm, outside diameter 2.03mm, inside diameter 1.63mm, wall thickness 0.2mm, annealed, 99.6+%
Titanium, rod, 500mm, diameter 6mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 2mm, as drawn, 99.99+%
Titanium, tube, 1000mm, outside diameter 9.5mm, inside diameter 8.2mm, wall thickness 0.65mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 10.3mm, inside diameter 8.7mm, wall thickness 0.8mm, annealed, 99.6+%