Merck
  • Zinc supplementation sustained normative neurodevelopment in a randomized, controlled trial of Peruvian infants aged 6-18 months.
Zinc supplementation sustained normative neurodevelopment in a randomized, controlled trial of Peruvian infants aged 6-18 months.
The Journal of nutrition (2014-05-23)
John Colombo, Nelly Zavaleta, Kathleen N Kannass, Fabiola Lazarte, Carla Albornoz, Leah L Kapa, Laura E Caulfield
ABSTRACT
A double-blind, randomized clinical trial was conducted to determine the effects of prevention of zinc deficiency on cognitive and sensorimotor development during infancy. At 6 mo of age, infants were randomly assigned to be administered a daily liquid supplement containing 10 mg/d of zinc (zinc sulfate), 10 mg/d of iron (ferrous sulfate), and 0.5 mg/d of copper (copper oxide), or an identical daily liquid supplement containing only 10 mg/d of iron and 0.5 mg/d of copper. Various controls were implemented to ensure adherence to the supplement protocol. A battery of developmental assessments was administered from 6 to 18 mo of age that included a visual habituation/recognition memory task augmented with heart rate at 6, 9, and 12 mo of age; the Bayley Scales of Infant Development, 2nd edition (BSID2) at 6, 12, and 18 mo; the A-not-B error task at 9 and 12 mo; and free-play attention tasks at 12 and 18 mo. Only infants supplemented with zinc had the normative decline in look duration from 6 to 12 mo during habituation and a normative decline in shifting between objects on free-play multiple-object attention tasks from 12 to 18 mo of age. The 2 groups did not differ on any of the psychophysiologic indices, the BSID2, or the A-not-B error task. The findings are consistent with zinc supplementation supporting a profile of normative information processing and active attentional profiles during the first 2 y of life. This trial was registered at clinicaltrials.gov as NCT00589264.
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Iron(II) sulfate heptahydrate, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Copper - O.F.H.C., tube, 1000mm, outside diameter 0.38mm, inside diameter 0.22mm, wall thickness 0.08mm, 99.95+%
Copper - O.F.H.C., wire reel, 20m, diameter 0.125mm, annealed, 99.95+%
Copper - O.F.H.C., rod, 100mm, diameter 76mm, as drawn, 99.95+%
Copper - O.F.H.C., wire reel, 2m, diameter 1.5mm, hard, 99.95+%