Merck
  • Glucose as a clean and renewable reductant in the Pd-nanoparticle-catalyzed reductive homocoupling of bromo- and chloroarenes in water.
Glucose as a clean and renewable reductant in the Pd-nanoparticle-catalyzed reductive homocoupling of bromo- and chloroarenes in water.
The Journal of organic chemistry (2010-05-14)
Antonio Monopoli, Vincenzo Calò, Francesco Ciminale, Pietro Cotugno, Carlo Angelici, Nicola Cioffi, Angelo Nacci
ABSTRACT
An efficient and highly sustainable Ullmann-type homocoupling of bromo- and chloroarenes, including the more challenging electron-rich chloroarenes (e.g., 4-chloroanisole), catalyzed by in situ generated Pd colloids, is carried out in aqueous medium under relatively mild conditions (temperatures ranging from 40 to 90 degrees C). Glucose is used as a clean and renewable reductant, while tetrabutylammonium hydroxide (TBAOH) acts as base, surfactant, and phase-transfer agent, creating a favorable environment for the catalyst. Pd nanoparticle sizes, morphology, and chemical composition are ascertained by TEM and XPS analyses.
Sigma-Aldrich
4-Chloroanisole, 99%