Merck
  • The role of the cemented all-polyethylene tibial component in total knee replacement: a 30-year patient follow-up and review of the literature.
The role of the cemented all-polyethylene tibial component in total knee replacement: a 30-year patient follow-up and review of the literature.
The Knee (2010-01-12)
Thomas J Blumenfeld, Richard D Scott
ABSTRACT
Use of an all-polyethylene tibial component in primary total knee arthroplasty remains an attractive option considering the reported durability of the construct, the lowered cost compared to modular metal-backed tibia, and the elimination of backside wear. The two major intra-operative disadvantages include the inability to alter the tibial component thickness after permanent implant placement and the inability to use varus-valgus constrained designs. The long-term disadvantage is the inability to perform a modular insert exchange should this be required. We report the 30-year outcome of a single patient using the duopatellar total knee replacement system. Based on a critical review of the literature we would recommend use in patients 80 years of age or older, consideration in patients 75 to 79 years, and possibly in younger yet less active patients. These three groups would be the least likely to require a modular tibial liner exchange in their lifetime.
Product Description
Polyethylene (LDPE), ERM®, certified reference material
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC