Merck
  • Ethiopian genetic diversity reveals linguistic stratification and complex influences on the Ethiopian gene pool.
Ethiopian genetic diversity reveals linguistic stratification and complex influences on the Ethiopian gene pool.
American journal of human genetics (2012-06-26)
Luca Pagani, Toomas Kivisild, Ayele Tarekegn, Rosemary Ekong, Chris Plaster, Irene Gallego Romero, Qasim Ayub, S Qasim Mehdi, Mark G Thomas, Donata Luiselli, Endashaw Bekele, Neil Bradman, David J Balding, Chris Tyler-Smith
ABSTRACT
Humans and their ancestors have traversed the Ethiopian landscape for millions of years, and present-day Ethiopians show great cultural, linguistic, and historical diversity, which makes them essential for understanding African variability and human origins. We genotyped 235 individuals from ten Ethiopian and two neighboring (South Sudanese and Somali) populations on an Illumina Omni 1M chip. Genotypes were compared with published data from several African and non-African populations. Principal-component and STRUCTURE-like analyses confirmed substantial genetic diversity both within and between populations, and revealed a match between genetic data and linguistic affiliation. Using comparisons with African and non-African reference samples in 40-SNP genomic windows, we identified "African" and "non-African" haplotypic components for each Ethiopian individual. The non-African component, which includes the SLC24A5 allele associated with light skin pigmentation in Europeans, may represent gene flow into Africa, which we estimate to have occurred ~3 thousand years ago (kya). The non-African component was found to be more similar to populations inhabiting the Levant rather than the Arabian Peninsula, but the principal route for the expansion out of Africa ~60 kya remains unresolved. Linkage-disequilibrium decay with genomic distance was less rapid in both the whole genome and the African component than in southern African samples, suggesting a less ancient history for Ethiopian populations.
Product Description
Genomiphi Hy Kit, Cytiva 25-6600-20, sufficient for 100 reactions
Genomiphi Hy Kit, Cytiva 25-6600-25, sufficient for 1000 reactions
GenomiPhi HY Kit, Cytiva 25-6600-22, sufficient for 25 reactions